missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-pyridinecarboxaldehyde, 97%, ACROS Organics™

Brand:  Acros Organics 442170050

Code : 29

Additional Details : CAS Number : 40273-47-0 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11946431

  • 242.00 € / 5g

Estimated Shipment: 07-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

40273-47-0
125.10
KLKQXQXSZCNWLZ-UHFFFAOYSA-N
2763003
FC1=C(C=O)C=CN=C1
C6H4FNO
MFCD06410686
3-fluoroisonicotinaldehyde, 3-fluoropyridine-4-carboxaldehyde, 3-fluoro-4-formylpyridine, 3-fluoro-4-pyridinecarboxaldehyde, 4-pyridinecarboxaldehyde, 3-fluoro, 3-fluoro-pyridine-4-carbaldehyde, 3-fluoro-4-pyridinecarbaldehyde, brettphospd, pubchem5137, acmc-20a1qb
3-fluoropyridine-4-carbaldehyde

Specifications

3-Fluoro-4-pyridinecarboxaldehyde
67°C
70.0°C (25.0 Torr)
40273-47-0
C6H4FNO
3-fluoroisonicotinaldehyde, 3-fluoropyridine-4-carboxaldehyde, 3-fluoro-4-formylpyridine, 3-fluoro-4-pyridinecarboxaldehyde, 4-pyridinecarboxaldehyde, 3-fluoro, 3-fluoro-pyridine-4-carbaldehyde, 3-fluoro-4-pyridinecarbaldehyde, brettphospd, pubchem5137, acmc-20a1qb
FC1=C(C=O)C=CN=C1
125.10
125.1
1.2550g/mL
1.255
5g
96% min. (GC)
MFCD06410686
KLKQXQXSZCNWLZ-UHFFFAOYSA-N
3-fluoropyridine-4-carbaldehyde
2763003
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Fluoro-4-pyridinecarboxaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates