missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Catalog Number p-4812946
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
161622-05-5
C8H3F4O2
207.10
MFCD00061293
NSGKIIGVPBTOBF-UHFFFAOYSA-M
3-fluoro-5-trifluoromethyl benzoic acid, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, 5-fluoro-3-trifluoromethyl benzoic acid, maybridge4_001933, acmc-1bwlq, intermediates-zcf02214, ksc496a7r, rarechem al bo 0634, buttpark 32\01-71
519222
3-fluoro-5-(trifluoromethyl)benzoic acid
[O-]C(=O)C1=CC(F)=CC(=C1)C(F)(F)F
This item is not returnable. View return policy
161622-05-5
C8H3F4O2
207.10
MFCD00061293
NSGKIIGVPBTOBF-UHFFFAOYSA-M
3-fluoro-5-trifluoromethyl benzoic acid, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, 5-fluoro-3-trifluoromethyl benzoic acid, maybridge4_001933, acmc-1bwlq, intermediates-zcf02214, ksc496a7r, rarechem al bo 0634, buttpark 32\01-71
519222
3-fluoro-5-(trifluoromethyl)benzoic acid
[O-]C(=O)C1=CC(F)=CC(=C1)C(F)(F)F
Melting Point 105.0°C to 106.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 208.11
Percent Purity 98%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzoic acid
compliance-icons
Product Identifier
  • 3-Fluoro-5-(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'