missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-2-one, 97%, Thermo Scientific™

Catalog Number p-3211921
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
1679-36-3
C6H8O
96.129
MFCD00041627
LTAPKZGQTMVYMX-UHFFFAOYSA-N
3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
137151
hex-3-yn-2-one
CCC#CC(=O)C
This item is not returnable. View return policy
1679-36-3
C6H8O
96.129
MFCD00041627
LTAPKZGQTMVYMX-UHFFFAOYSA-N
3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
137151
hex-3-yn-2-one
CCC#CC(=O)C
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.8780g/mL
Boiling Point 68.0°C (50.0 mmHg)
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3CH2C≡CCOCH3
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.878
Formula Weight 96.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Hexyn-2-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'