missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methyl-2-butanone, 92%, Thermo Scientific Chemicals

Catalog Number p-146056
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
115-22-0
C5H10O2
102.13
MFCD00004460
BNDRWEVUODOUDW-UHFFFAOYSA-N
3-hydroxy-3-methyl-2-butanone, 3-methylacetoin, methylacetoin, 2-butanone, 3-hydroxy-3-methyl, dimethylacetylcarbinol, 2-hydroxy-2-methyl-3-butanone, 2-methyl-2-hydroxybutan-3-one, 1-hydroxy-1-methylethyl methyl ketone, 3-hydroxy-3-methyl-butan-2-one, 3-hydroxy-3-ethylbutanone
8261
3-hydroxy-3-methylbutan-2-one
CC(=O)C(C)(C)O
This item is not returnable. View return policy
115-22-0
C5H10O2
102.13
MFCD00004460
BNDRWEVUODOUDW-UHFFFAOYSA-N
3-hydroxy-3-methyl-2-butanone, 3-methylacetoin, methylacetoin, 2-butanone, 3-hydroxy-3-methyl, dimethylacetylcarbinol, 2-hydroxy-2-methyl-3-butanone, 2-methyl-2-hydroxybutan-3-one, 1-hydroxy-1-methylethyl methyl ketone, 3-hydroxy-3-methyl-butan-2-one, 3-hydroxy-3-ethylbutanone
8261
3-hydroxy-3-methylbutan-2-one
CC(=O)C(C)(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 90.0
CAS Max % 100.0
Color Colorless to Yellow or Red
Density 0.9700g/mL
Boiling Point 140°C to 141°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C(OH)COCH3
Refractive Index 1.4145 to 1.4165
Quantity 5 g
Beilstein 01, 832
Specific Gravity 0.97
Formula Weight 102.13
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methyl-2-butanone, 92%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-3-methyl-2-butanone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'