Learn More
3-Isocyanato-5-methyl-2-phenylfuran, 97%, Maybridge™
Brand: Maybridge CC49706CB
Code : 31
Additional Details : CAS Number : 898289-18-4 Weight : 0.05025kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
898289-18-4 | |
199.209 | |
GDLFJANXKGIZKA-UHFFFAOYSA-N | |
18525836 | |
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O |
C12H9NO2 | |
MFCD09025851 | |
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl | |
3-isocyanato-5-methyl-2-phenylfuran |
Specifications
3-Isocyanato-5-methyl-2-phenylfuran | |
250mg | |
96.5 | |
97% | |
MFCD09025851 | |
GDLFJANXKGIZKA-UHFFFAOYSA-N | |
3-isocyanato-5-methyl-2-phenylfuran | |
18525836 | |
97% |
Amber Glass Bottle | |
898289-18-4 | |
100.0 | |
C12H9NO2 | |
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl | |
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O | |
199.209 | |
199.21 |