missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number 11966111
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
603122-40-3
C15H21BO5
292.14
This item is not returnable. View return policy
603122-40-3
C15H21BO5
292.14

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 71°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 292.14
Percent Purity 95%
Chemical Name or Material 3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Methoxy-4-(methoxycarbonyl)phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'