missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-butyne, 96%, Thermo Scientific Chemicals
66.60 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Store in cool place. Light and heat sensitive. Incompatible with strong oxidizing agents, acids and bases.
3-Methyl-1-butyne is a terminal alkyne that under goes hydrosilylation reaction using phenylsilane. 3-Methyl-1-butyne is used as an intermediate for pharmaceuticals, in cosmetics and in chemical research.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Store in cool place. Light and heat sensitive. Incompatible with strong oxidizing agents, acids and bases.
Specifications
Density | 0.666 |
Boiling Point | 30°C |
Flash Point | −30°C (−22°F) |
Linear Formula | (CH3)2CHC≡CH |
Refractive Index | 1.374 |
Quantity | 5 g |
UN Number | UN3295 |
Solubility Information | Miscible with alcohol. Immiscible with water. |
Formula Weight | 68.12 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'