missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-buten-1-ol, 97%, Thermo Scientific Chemicals

Catalog Number p-160785
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
763-32-6
C5H10O
86.13
MFCD00002933
CPJRRXSHAYUTGL-UHFFFAOYSA-N
3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
12988
CHEBI:62898
3-methylbut-3-en-1-ol
CC(=C)CCO
This item is not returnable. View return policy
763-32-6
C5H10O
86.13
MFCD00002933
CPJRRXSHAYUTGL-UHFFFAOYSA-N
3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
12988
CHEBI:62898
3-methylbut-3-en-1-ol
CC(=C)CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 132°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CH2CH2OH
Refractive Index 1.4327 to 1.4347
Quantity 100 mL
Specific Gravity 0.85
Solubility Information Solubility in water: 170g/L (20 c)
Formula Weight 86.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-buten-1-ol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methyl-3-buten-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'