missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-methoxybutanol, 99%, Thermo Scientific Chemicals

Catalog Number 10006043
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
56539-66-3
MFCD00044771
MFKRHJVUCZRDTF-UHFFFAOYSA-N
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
62118
3-methoxy-3-methylbutan-1-ol
CC(C)(CCO)OC
This item is not returnable. View return policy
56539-66-3
MFCD00044771
MFKRHJVUCZRDTF-UHFFFAOYSA-N
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389
62118
3-methoxy-3-methylbutan-1-ol
CC(C)(CCO)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless
Density 0.9260g/mL
Boiling Point 174.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H14O2
Linear Formula (CH3)2C(OCH3)CH2CH2OH
Refractive Index 1.4260 to 1.4280
Quantity 1 L
Specific Gravity 0.926
Molecular Weight (g/mol) 118.176
Formula Weight 118.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-methoxybutanol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-3-methoxybutanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'