missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methyl-N-(3,5-dimethylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59470.03

Code : NEW

Additional Details : CAS Number : 346720-25-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415808

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Methyl-N-(3,5-dimethylphenyl)benzamide
346720-25-0
239.32
MFCD01034055

Specifications

3-Methyl-N-(3,5-dimethylphenyl)benzamide
Solid
239.32
1g
346720-25-0
97%
MFCD01034055
Videos
SDS