missing translation for 'onlineSavingsMsg'
Learn More

3-(N-BOC-aminomethyl)-5-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog Number 11946461
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
943722-24-5
C11H15BrN2O2
287.16
VZPOHASZICQBDX-UHFFFAOYSA-N
3-n-boc-aminomethyl-5-bromopyridine, tert-butyl 5-bromopyridin-3-yl methylcarbamate, tert-butyl 5-bromopyridin-3-yl methyl carbamate, tert-butyl n-5-bromopyridin-3-yl methyl carbamate, 3-bromo-5-tert-butoxycarbonyl amino methyl pyridine, 3-boc-aminomethyl-5-bromopyridine, 5-bromo-3-boc-amino methyl pyridine, 5-bromo-3-n-tert-butyl carbamatomethylpyridine, 3-aminomethyl-5-bromopyridine, 3-boc protected
46739290
tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC(=CN=C1)Br
This item is not returnable. View return policy
943722-24-5
C11H15BrN2O2
287.16
VZPOHASZICQBDX-UHFFFAOYSA-N
3-n-boc-aminomethyl-5-bromopyridine, tert-butyl 5-bromopyridin-3-yl methylcarbamate, tert-butyl 5-bromopyridin-3-yl methyl carbamate, tert-butyl n-5-bromopyridin-3-yl methyl carbamate, 3-bromo-5-tert-butoxycarbonyl amino methyl pyridine, 3-boc-aminomethyl-5-bromopyridine, 5-bromo-3-boc-amino methyl pyridine, 5-bromo-3-n-tert-butyl carbamatomethylpyridine, 3-aminomethyl-5-bromopyridine, 3-boc protected
46739290
tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC(=CN=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 84.5°C to 86°C
Assay Percent Range 96% min. (GC)
Formula Weight 287.16
Percent Purity 97%
Quantity 5 g
Chemical Name or Material 3-(N-BOC-aminomethyl)-5-bromopyridine
compliance-icons
Product Identifier
  • 3-(N-BOC-Aminomethyl)-5-bromopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'