missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyltoluene, 95%, Thermo Scientific Chemicals

Catalog Number 10003620
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
643-93-6
C13H12
168.24
MFCD00008533
NPDIDUXTRAITDE-UHFFFAOYSA-N
3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
12564
1-methyl-3-phenylbenzene
CC1=CC=CC(=C1)C2=CC=CC=C2
This item is not returnable. View return policy
643-93-6
C13H12
168.24
MFCD00008533
NPDIDUXTRAITDE-UHFFFAOYSA-N
3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
12564
1-methyl-3-phenylbenzene
CC1=CC=CC(=C1)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 4°C to 5°C
Color Yellow
Density 1.0100g/mL
Boiling Point 272°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4CH3
Refractive Index 1.6010 to 1.605
Quantity 1 g
Specific Gravity 1.01
Formula Weight 168.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Phenyltoluene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'