missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Piperidin-1-ylmethyl)benzoic acid hydrochloride hemihydrate, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 264.76 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Piperidinomethyl)benzoic acid hydrochloride 0.5 hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'