missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperidinomethyl)benzaldehyde, ≥97%, Maybridge™

Brand:  Maybridge CC45204DA

Code : 31

Additional Details : Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 10027852

  • 86.56 € / 1g

Estimated Shipment: 22-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

471930-01-5
203.285
UJDTVJKIOWAUNQ-UHFFFAOYSA-N
7164643
C1CCN(CC1)CC2=CC=CC(=C2)C=O
C13H17NO
MFCD07782418
3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
3-(piperidin-1-ylmethyl)benzaldehyde

Specifications

3-(Piperidinomethyl)benzaldehyde
1g
96.5
97%
MFCD07782418
UJDTVJKIOWAUNQ-UHFFFAOYSA-N
3-(piperidin-1-ylmethyl)benzaldehyde
7164643
97%
Amber Glass Bottle
471930-01-5
100.0
C13H17NO
3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
C1CCN(CC1)CC2=CC=CC(=C2)C=O
203.285
203.28
Videos
SDS Documents
Certificates