missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridazinecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4380109
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2164-61-6
C5H4N2O2
124.1
RUUOPSRRIKJHNH-UHFFFAOYSA-N
3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid;
269369
pyridazine-3-carboxylic acid
C1=CC(=NN=C1)C(=O)O
This item is not returnable. View return policy
2164-61-6
C5H4N2O2
124.1
RUUOPSRRIKJHNH-UHFFFAOYSA-N
3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid;
269369
pyridazine-3-carboxylic acid
C1=CC(=NN=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190.8°C to 194.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material 3-Pyridazinecarboxylic acid
compliance-icons
Product Identifier
  • Pyridazine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'