Learn More
3-Pyridinecarboxaldehyde, 98%, ACROS Organics™
Brand: Acros Organics 131835000
Code : 29
Additional Details : CAS Number : 500-22-1 Weight : 0.55000kg Transport : UN number : 3265 Chem class : 8 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 500g |
Chemical Identifiers
500-22-1 | |
107.11 | |
3-pyridinecarboxaldehyde, nicotinaldehyde, 3-pyridinaldehyde, pyridine-3-carboxaldehyde, 3-pyridylcarboxaldehyde, 3-pyridylaldehyde, nicotinealdehyde, 3-formylpyridine, pyridine-3-aldehyde, nicotinic aldehyde | |
CHEBI:28345 | |
C1=CC(=CN=C1)C=O |
C6H5NO | |
QJZUKDFHGGYHMC-UHFFFAOYSA-N | |
10371 | |
pyridine-3-carbaldehyde |
Specifications
3-Pyridinecarboxaldehyde | |
84°C | |
Glass bottle | |
1.142 | |
Yellow to Brown | |
500-22-1 | |
C6H5NO | |
21,I,288 | |
Solubility in water: miscible. Other solubilities: soluble in most organic solvents | |
C1=CC(=CN=C1)C=O | |
107.11 | |
CHEBI:28345 | |
Liquid |
1.1420g/mL | |
Authentic | |
1.5480 to 1.5500 | |
95.0°C to 97.0°C (15.0mmHg) | |
500g | |
97.5% min. (GC) | |
(C5H4N)COH | |
3-pyridinecarboxaldehyde, nicotinaldehyde, 3-pyridinaldehyde, pyridine-3-carboxaldehyde, 3-pyridylcarboxaldehyde, 3-pyridylaldehyde, nicotinealdehyde, 3-formylpyridine, pyridine-3-aldehyde, nicotinic aldehyde | |
QJZUKDFHGGYHMC-UHFFFAOYSA-N | |
pyridine-3-carbaldehyde | |
10371 | |
107.11 | |
98% |
Safety and Handling
- 3-Pyridinecarboxaldehyde
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .