missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-2-ylaniline, 97%, Thermo Scientific™

Catalog Number 10012074
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
69491-56-1
C10H9N3
171.203
MFCD09702385
GOCBKTKLGFLECW-UHFFFAOYSA-N
3-pyrimidin-2-yl aniline, 3-pyrimidin-2-ylphenylamine, 2-3-aminophenyl pyrimidine, 2-3-aminophenyl-1,3-diazine, benzenamine, 3-2-pyrimidinyl
12456169
3-pyrimidin-2-ylaniline
C1=CC(=CC(=C1)N)C2=NC=CC=N2
This item is not returnable. View return policy
69491-56-1
C10H9N3
171.203
MFCD09702385
GOCBKTKLGFLECW-UHFFFAOYSA-N
3-pyrimidin-2-yl aniline, 3-pyrimidin-2-ylphenylamine, 2-3-aminophenyl pyrimidine, 2-3-aminophenyl-1,3-diazine, benzenamine, 3-2-pyrimidinyl
12456169
3-pyrimidin-2-ylaniline
C1=CC(=CC(=C1)N)C2=NC=CC=N2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 171.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-2-ylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'