missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylaniline, ≥97%, Maybridge™

Brand:  Maybridge CC41714DE

Code : 31

Additional Details : Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 10011844

  • 260.00 € / 5g

Estimated Shipment
01-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

69491-59-4
171.203
DZEIKJMNXHOFHL-UHFFFAOYSA-N
7162049
C1=CC(=CC(=C1)N)C2=CN=CN=C2
C10H9N3
MFCD06802522
3-pyrimidin-5-yl aniline, 3-5-pyrimidinyl aniline, benzenamine,3-5-pyrimidinyl, 3-5-pyrimidyl aniline, 3-5-pyrimidyl-aniline, 3-pyrimidin-5-ylphenylamine, 3-pyrimidin-5-yl-phenylamine, 3-pyrimidin-5-yl phenylamine, benzenamine, 3-5-pyrimidinyl
3-pyrimidin-5-ylaniline

Specifications

3-Pyrimidin-5-ylaniline
69491-59-4
100.0
C10H9N3
3-pyrimidin-5-yl aniline, 3-5-pyrimidinyl aniline, benzenamine,3-5-pyrimidinyl, 3-5-pyrimidyl aniline, 3-5-pyrimidyl-aniline, 3-pyrimidin-5-ylphenylamine, 3-pyrimidin-5-yl-phenylamine, 3-pyrimidin-5-yl phenylamine, benzenamine, 3-5-pyrimidinyl
C1=CC(=CC(=C1)N)C2=CN=CN=C2
171.203
171.2
5g
96.5
97%
MFCD06802522
DZEIKJMNXHOFHL-UHFFFAOYSA-N
3-pyrimidin-5-ylaniline
7162049
97%
Videos
SDS Documents