missing translation for 'onlineSavingsMsg'
Learn More

(3-Pyrrolidin-1-ylphenyl)methylamine, ≥90%, Thermo Scientific™

Catalog Number p-3176043
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175696-70-5
C11H16N2
176.26
MFCD07772827
BXTWTBYGOKOWQX-UHFFFAOYSA-N
3-pyrrolidin-1-ylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidinobenzylamine, 1-3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidin-1-ylphenyl methanamine, 3-pyrrolizinobenzylamine, 3-pyrrolidin-1-yl benzylamine, 3-pyrrolidinylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methylamine
7162061
(3-pyrrolidin-1-ylphenyl)methanamine
NCC1=CC(=CC=C1)N1CCCC1
This item is not returnable. View return policy
175696-70-5
C11H16N2
176.26
MFCD07772827
BXTWTBYGOKOWQX-UHFFFAOYSA-N
3-pyrrolidin-1-ylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidinobenzylamine, 1-3-pyrrolidin-1-yl phenyl methanamine, 3-pyrrolidin-1-ylphenyl methanamine, 3-pyrrolizinobenzylamine, 3-pyrrolidin-1-yl benzylamine, 3-pyrrolidinylphenyl methylamine, 3-pyrrolidin-1-yl phenyl methylamine
7162061
(3-pyrrolidin-1-ylphenyl)methanamine
NCC1=CC(=CC=C1)N1CCCC1
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.26
Percent Purity 90%
Chemical Name or Material (3-Pyrrolidin-1-ylphenyl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'