missing translation for 'onlineSavingsMsg'
Learn More

Ismatec 3-Stop Tubing, Fluran F-5500-A

Specially formulated elastomer for use with strong acidic and basic solutions.

Brand:  Ismatec SC0263

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10012862

  • 195.00 € / Pack of 12


Estimated Shipment
16-02-2021
Add to basket

Description

Description

  • For use with pumps that incorporate cassettes into the pumphead design.
  • Color-coded stops.
  • Specially formulated elastomer for use with strong acid and basic solutions.
  • Very low gas permeability.
  • The choice material for applications where fluids can be transferred without being contaminated by atmospheric gases.
  • Low gas permeability and relative strength make it a material of choice in vacuum-based applications.
Specifications

Specifications

Black
Yellow-Blue
Fluran F-5500-A
For use with strong acidic and basic solutions, pumps with cassettes in pumphead design.
12/Pk
1.52 mm
204°C
1.52 mm
400 mm
&minus31°C
Acids: Excellent, Alkaline solutions: Excellent, Solvents: Limited, Pressure: Not rec., Vacuum: Good, Viscous media: Good, Sterile media: Fair
Videos
SDS
Documents