missing translation for 'onlineSavingsMsg'
Learn More

3-Thienylboronic acid, 97%, Maybridge™

Catalog Number 10001744
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6165-69-1
C4H5BO2S
127.95
MFCD00151851
QNMBSXGYAQZCTN-UHFFFAOYSA-N
3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
581760
(thiophen-3-yl)boronic acid
OB(O)C1=CSC=C1
This item is not returnable. View return policy
6165-69-1
C4H5BO2S
127.95
MFCD00151851
QNMBSXGYAQZCTN-UHFFFAOYSA-N
3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid, thiophen-3-yl-3-boronic acid, 3-thienyl boronic acid, 3-thiophene boronic acid, thiophen-3-yl-boranediol, boronic acid, 3-thienyl
581760
(thiophen-3-yl)boronic acid
OB(O)C1=CSC=C1
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 127.96
Percent Purity 97%
Chemical Name or Material 3-Thienylboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'