missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(Trifluoromethyl)benzamidoxime, 97%

Brand:  Alfa Aesar™ H58744.04

Code : NEW

Additional Details : CAS Number : 40067-80-9 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 15414968

  • 156.88 € / 2g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(Trifluoromethyl)benzamidoxime
40067-80-9
204.15
MFCD02647312

Specifications

3-(Trifluoromethyl)benzamidoxime
82° to 88°C
40067-80-9
97%
MFCD02647312
Pale brown
2g
Solid
204.15
Videos
SDS