missing translation for 'onlineSavingsMsg'
Learn More

3-(Triphenylphosphoranylidene)tetrahydropyran-2-one, 97%, ACROS Organics™

Brand:  Acros Organics 440980010

Code : 29

Additional Details : CAS Number : 2234-82-4 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11956021

  • 142.00 € / 1g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

218282-19-0
360.39
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one
C23H21O2P
PZGUKMFSICDTKX-UHFFFAOYSA-N
10594674
C1CC(=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)OC1

Specifications

3-(Triphenylphosphoranylidene)tetrahydropyran-2-one
228.0°C to 230.0°C
218282-19-0
C23H21O2P
PZGUKMFSICDTKX-UHFFFAOYSA-N
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one
10594674
97%
Glass bottle
1g
96% min. (HPLC)
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one
C1CC(=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)OC1
360.39
360.39
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-(3-(Triphenylphosphoranylidene)tetrahydropyran-2-one

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates