missing translation for 'onlineSavingsMsg'
Learn More

3M™ Hooded Anti-Static Protective Coveralls 4565 Series

Catalog Number p-4361078
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Comfortable coveralls made with high-performance fabric offer advanced quality and protection in challenging conditions such as exposure to hazardous dusts and particulates.

  • Type 4/5/6 Coverall designed to offer a very good barrier to hazardous particles and limited liquid splash and spray
  • Made of bicomponent nonwoven with ultra-low linting PE laminate for a noticeably comfortable drape
  • 3 panel hood, 2-way zip and storm flap with sealable tape with elasticated back and ankles to reduce the risk of skin exposure
  • Taped seams across the suit for increased hold out and strength reducing the risk of tearing and exposure
  • Protection against EN 14126 Biological agents
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out Til class 2
Product Type Cleanroom Coveralls
Size Medium
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Knit
Closure Type 2-way Zipper with Sealed Storm Flap
Seam Type Taped
Sterility Non-sterile
Coating Anti-static
Certifications/Compliance ASTM F1670, EN 14126:2003, EN 1149-1:2006, EN 13034, EN-ISO 13982-30
Description Polyethylene and Polyester/Polyethylene laminate suit, Metal/Nylon/Polyester Braid zipper, Neoprene Rubber elastic, Polyester/Cotton thread, Polyester cuffs, Polypropylene red tape
Size (English) Chest 36 to 39 in.
Size (English) Inseam 30 in.
Size (English) Waist 50 in.
Size (Metric) Chest 91.44 to 99.06 cm
Size (Metric) Inseam 76.2 cm
Size (Metric) Waist 127 cm
For Use With (Application) Paint Spraying, Industrial and Pharmaceutical Manufacturing, Asbestos Inspection and Handling, Healthcare
Height (English) 5.5 to 5.75 ft.
Height (Metric) 167 to 176 cm
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'