Learn More
3M™ Disposable Protective Overboot Cover 441
The 3M™ Disposable boot covers are designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.
Brand: 3M™ 7000089719
33.70 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.79200kg
Description
The 3M™ Overboot Covers 440 are a reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Offered as an accessory, the overboots are made from a microporous laminate material with an anti-static treatment.- Covers are designed with ties and elastic at the calf to help ensure a good fit
- Distinctive unisex design
- Integrated sideshields with adjustable lens angle and temple lengths
- Vertically vented browgard minimizes fogging
Specifications
Universal | |
White | |
Elastic | |
Dry particulates |
Microporous fabric | |
Single-use | |
Ideal to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts and for assembly, cleaning, machine operations | |
Boot Covers |