Learn More
3M™ Reusable Respirator Accessory, Filter Adapter 603
Used with 3M™ Particulate Filter 5N11 or 3M™ Particulate Filter 5P71 and 3M™ Filter Retainer 501.
Brand: 3M™ 7000029662
Code : NEW
Additional Details : Weight : 1.00000kg
Description
The 3M™ Filter Adapter 603 enables the 5N11 and 5P71 N95 and P95 prefilters to be directly attached to a 3M™ Full or Half Facepiece 6000, 7000 or FF-400 Series Respirator. The filter adapter 603 may be used multiple times. It provides cost-effective filtering solution when multiple filters are needed for 3M elastomeric facepieces.- Approved 3M system component
- Designed to attach 3M Filters 5N11 and 5P71 N95 and P95 with 3M 501 Filter Retainer
- Component works with 3M Full or Half Facepieces 6000, 7000, or FF-400 Series Respirators (sold separately)
- Filter adapter may be used multiple times for convenience
- Cost effective particle filter solution
- 3M Catalog# 603
Specifications
Filter Cartridge | |
Reusable Respirator | |
603 | |
16/Cs. |