missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog Number 10423573
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
160068-88-2
C10H11BO3
190.005
MFCD04039026
AMQBUMNOSGJNQL-UHFFFAOYSA-N
4-1,3,2-dioxaborinan-2-yl benzaldehyde, 4-formylbenzeneboronic acid, propane-1,3-diol cyclic ester, 4-formylbenzeneboronic acid,propane-1,3-diol cyclic ester, 4-1,3,2-dioxaboran-2-yl benzaldehyde, 2-4-formylphenyl-1,3,2-dioxaborinane
2794711
4-(1,3,2-dioxaborinan-2-yl)benzaldehyde
B1(OCCCO1)C2=CC=C(C=C2)C=O
This item is not returnable. View return policy
160068-88-2
C10H11BO3
190.005
MFCD04039026
AMQBUMNOSGJNQL-UHFFFAOYSA-N
4-1,3,2-dioxaborinan-2-yl benzaldehyde, 4-formylbenzeneboronic acid, propane-1,3-diol cyclic ester, 4-formylbenzeneboronic acid,propane-1,3-diol cyclic ester, 4-1,3,2-dioxaboran-2-yl benzaldehyde, 2-4-formylphenyl-1,3,2-dioxaborinane
2794711
4-(1,3,2-dioxaborinan-2-yl)benzaldehyde
B1(OCCCO1)C2=CC=C(C=C2)C=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 190
Percent Purity 97%
Chemical Name or Material 4-(1, 3, 2-Dioxaborinan-2-yl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'