missing translation for 'onlineSavingsMsg'
Learn More

4-{[2-(Dimethylamino)ethyl]carbamoyl}phenylboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number 11946091
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
832114-11-1
C17H27BN2O3
318.23
KTDBEJMIGCLCSU-UHFFFAOYSA-N
n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-n',n'-dimethylamino ethyl benzamide-4-boronic acid, pinacol ester, n-2-dimethylamino ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylamino ethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-dimethylamino ethylcarbamoyl phenylboronic acid, pinacol ester, amtb351, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,, 4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester, 4-n-2-dimethylamino ethyl carbamoyl benzeneboronic acid pinacol ester
2758503
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C
This item is not returnable. View return policy
832114-11-1
C17H27BN2O3
318.23
KTDBEJMIGCLCSU-UHFFFAOYSA-N
n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-n',n'-dimethylamino ethyl benzamide-4-boronic acid, pinacol ester, n-2-dimethylamino ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylamino ethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-dimethylamino ethylcarbamoyl phenylboronic acid, pinacol ester, amtb351, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,, 4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester, 4-n-2-dimethylamino ethyl carbamoyl benzeneboronic acid pinacol ester
2758503
N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C
Melting Point 107°C to 110°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 318.23
Percent Purity 95%
Chemical Name or Material 4-{[2-(Dimethylamino)ethyl]carbamoyl}phenylboronic acid pinacol ester
compliance-icons compliance-icons
Product Identifier
  • 4-{¢2-(Dimethylamino)ethyl!carbamoyl}phenylboronic acid pinacol ester
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'