missing translation for 'onlineSavingsMsg'
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-2690476
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
171364-82-2
C13H16BNO2
229.09
MFCD03093897
HOPDTPGXBZCBNP-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyanophenylboronic acid pinacol ester, 4-cyanobenzeneboronic acid pinacol ester, 4-cyanophenylboronic acid, pinacol ester, benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-cyanobenzeneboronic acid, pinacol ester, 2-4-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 4-cyanophenylboronicacidpinacolester
2734625
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
171364-82-2
C13H16BNO2
229.09
MFCD03093897
HOPDTPGXBZCBNP-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyanophenylboronic acid pinacol ester, 4-cyanobenzeneboronic acid pinacol ester, 4-cyanophenylboronic acid, pinacol ester, benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-cyanobenzeneboronic acid, pinacol ester, 2-4-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 4-cyanophenylboronicacidpinacolester
2734625
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 94°C to 99°C
Color Orange to Yellow or Tan
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 229.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'