missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-chlorophenyl)sulfonyl]-3-methylthiophene-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10425512
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175137-65-2
C12H9ClO4S2
316.77
MFCD00067886
WZQQSJVPCVCQKE-UHFFFAOYSA-N
4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorobenzenesulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chloro-benzenesulfonyl-3-methyl-thiophene-2-carboxylic acid, maybridge1_004536, 4-4-chlorophenylsulfonyl-3-methylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 4-4-chlorophenyl sulfonyl-3-methyl, 4-4-chlorobenzene-1-sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methyl-thiophene-2-carboxylic acid
2778453
4-(4-chlorophenyl)sulfonyl-3-methylthiophene-2-carboxylic acid
CC1=C(SC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)C(=O)O
This item is not returnable. View return policy
175137-65-2
C12H9ClO4S2
316.77
MFCD00067886
WZQQSJVPCVCQKE-UHFFFAOYSA-N
4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorobenzenesulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chloro-benzenesulfonyl-3-methyl-thiophene-2-carboxylic acid, maybridge1_004536, 4-4-chlorophenylsulfonyl-3-methylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 4-4-chlorophenyl sulfonyl-3-methyl, 4-4-chlorobenzene-1-sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methyl-thiophene-2-carboxylic acid
2778453
4-(4-chlorophenyl)sulfonyl-3-methylthiophene-2-carboxylic acid
CC1=C(SC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 316.78
Percent Purity 97%
Chemical Name or Material 4-[(4-chlorophenyl)sulfonyl]-3-methylthiophene-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'