missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-chlorophenyl)sulfonyl]-3-methylthiophene-2-carboxylic acid, 97%, Maybridge

Brand:  Maybridge KM01338DA

Code : 31

Additional Details : CAS Number : 175137-65-2 Weight : 0.05100kg

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10425322

  • 57.40 € / 1g


Estimated Shipment
28-03-2019
Add to basket

Specifications

Chemical Identifiers

175137-65-2
316.77
WZQQSJVPCVCQKE-UHFFFAOYSA-N
2778453
CC1=C(SC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)C(=O)O
C12H9ClO4S2
MFCD00067886
4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorobenzenesulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chloro-benzenesulfonyl-3-methyl-thiophene-2-carboxylic acid, maybridge1_004536, 4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenylsulfonyl-3-methylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 4-4-chlorophenyl sulfonyl-3-methyl, 4-4-chlorobenzene-1-sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methyl-thiophene-2-carboxylic acid
4-(4-chlorophenyl)sulfonyl-3-methylthiophene-2-carboxylic acid

Specifications

4-[(4-chlorophenyl)sulfonyl]-3-methylthiophene-2-carboxylic acid
1g
97.0
C12H9ClO4S2
4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorobenzenesulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chloro-benzenesulfonyl-3-methyl-thiophene-2-carboxylic acid, maybridge1_004536, 4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenylsulfonyl-3-methylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 4-4-chlorophenyl sulfonyl-3-methyl, 4-4-chlorobenzene-1-sulfonyl-3-methylthiophene-2-carboxylic acid, 4-4-chlorophenyl sulfonyl-3-methyl-thiophene-2-carboxylic acid
CC1=C(SC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)C(=O)O
316.77
316.78
Amber Glass Bottle
175137-65-2
97%
MFCD00067886
WZQQSJVPCVCQKE-UHFFFAOYSA-N
4-(4-chlorophenyl)sulfonyl-3-methylthiophene-2-carboxylic acid
2778453
97%
Videos
SDS