missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dichlorobenzophenone, 99%, Thermo Scientific Chemicals
25.93 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.
Solubility
Soluble in hot acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Avoid strong oxidizing agents.
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.
Solubility
Soluble in hot acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Avoid strong oxidizing agents.
Specifications
Melting Point | 144°C to 146°C |
Boiling Point | 352°C to 354°C |
Quantity | 25 g |
Beilstein | 643345 |
Solubility Information | Soluble in hot acetone. |
Formula Weight | 251.11 |
Percent Purity | 99% |
Chemical Name or Material | 4,4'-Dichlorobenzophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'