missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethylbenzophenone, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4,4'-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
4,4′-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Melting Point | 92°C to 96°C |
Boiling Point | 200°C (17 mmHg) |
Quantity | 5 g |
Beilstein | 1240527 |
Solubility Information | Insoluble in water. |
Formula Weight | 210.28 |
Percent Purity | ≥98% |
Chemical Name or Material | 4,4'-Dimethylbenzophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'