missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dipyridyl, 98%, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4510397
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
553-26-4
C10H8N2
156.19
MFCD00006416
MWVTWFVJZLCBMC-UHFFFAOYSA-N
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
11107
CHEBI:30985
4-pyridin-4-ylpyridine
C1=CC(=CC=N1)C1=CC=NC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
553-26-4
C10H8N2
156.19
MFCD00006416
MWVTWFVJZLCBMC-UHFFFAOYSA-N
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
11107
CHEBI:30985
4-pyridin-4-ylpyridine
C1=CC(=CC=N1)C1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 110.0°C to 114.0°C
Color White to Beige
Boiling Point 305.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)-(C5H4N)
Quantity 5 g
Beilstein 23,2
Fieser 16,25
Merck Index 15,3389
Solubility Information Solubility in water: 4,5g/L (25°C). Other solubilities: freely soluble in alcohol, benzene, chloroform,, ether,
Formula Weight 156.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dipyridyl
Show More Show Less
compliance-icons
Product Identifier
  • 4,4`-Dipyridyl, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'