missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4'-(4-Fluoro-2-methylphenylsulfamoyl)acetanilide, 97%

Brand:  Alfa Aesar™ H60636.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475906

  • 646.00 € / 1g


Estimated Shipment date
from Supplier 20-06-2019
Add to basket

Specifications

Chemical Identifiers

C15H15FN2O3S
MFCD05094075

Specifications

4'-(4-Fluoro-2-methylphenylsulfamoyl)acetanilide
C15H15FN2O3S
322.35
1g
MFCD05094075
97%
Videos
SDS