Learn More
4-(4-Methylpiperazino)aniline, 97%, Maybridge™
Brand: Maybridge SS00001DA
Code : 31
Additional Details : CAS Number : 16153-81-4 Weight : 0.05100kg Transport : UN number : 2923 Chem class : 8 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
16153-81-4 | |
191.278 | |
MOZNZNKHRXRLLF-UHFFFAOYSA-N | |
737253 | |
CN1CCN(CC1)C2=CC=C(C=C2)N |
C11H17N3 | |
MFCD00172703 | |
4-4-methylpiperazino aniline, 4-4-methylpiperazin-1-yl aniline, 4-methyl-1-4-aminophenyl piperazine, 4-4-methyl-1-piperazinyl aniline, 4-4-methylpiperazin-1-yl phenylamine, 1-4-aminophenyl-4-methylpiperazine, 4-4-methyl-piperazin-1-yl-phenylamine, 4-4-methylpiperazin-1-yl benzenamine, benzenamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenylamine | |
4-(4-methylpiperazin-1-yl)aniline |
Specifications
4-(4-Methylpiperazino)aniline | |
1g | |
97.0 | |
C11H17N3 | |
4-4-methylpiperazino aniline, 4-4-methylpiperazin-1-yl aniline, 4-methyl-1-4-aminophenyl piperazine, 4-4-methyl-1-piperazinyl aniline, 4-4-methylpiperazin-1-yl phenylamine, 1-4-aminophenyl-4-methylpiperazine, 4-4-methyl-piperazin-1-yl-phenylamine, 4-4-methylpiperazin-1-yl benzenamine, benzenamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenylamine | |
CN1CCN(CC1)C2=CC=C(C=C2)N | |
191.278 | |
191.28 |
Amber glass bottle | |
16153-81-4 | |
97% | |
MFCD00172703 | |
MOZNZNKHRXRLLF-UHFFFAOYSA-N | |
4-(4-methylpiperazin-1-yl)aniline | |
737253 | |
97% |