Learn More
4,5-Difluoro-2-nitrobenzoic acid, 99%, ACROS Organics™
Brand: Acros Organics 442670010
Code : 29
Additional Details : CAS Number : 20372-63-8 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
20372-63-8 | |
203.1 | |
4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 3,4-difluoro-6-nitrobenzoic acid, 2-nitro-4,5-difluorobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, benzoic acid, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-two fluoro-2-nitrobenzoic acid, pubchem4947, acmc-1cegm | |
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O |
C7H3F2NO4 | |
HGGRAOYTQNFGGN-UHFFFAOYSA-N | |
4219630 |
Specifications
4, 5-Difluoro-2-nitrobenzoic acid | |
166.0°C to 167.0°C | |
20372-63-8 | |
C7H3F2NO4 | |
HGGRAOYTQNFGGN-UHFFFAOYSA-N | |
203.1 | |
203.1 |
Glass Bottle | |
1g | |
98.5% min. (HPLC) | |
4,5-difluoro-2-nitrobenzoic acid, 4,5-difluoro-2-nitrobenzoicacid, 3,4-difluoro-6-nitrobenzoic acid, 2-nitro-4,5-difluorobenzoic acid, 4,5-difluoro-2-nitro-benzoic acid, benzoic acid, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitrobenzenecarboxylic acid, 4,5-two fluoro-2-nitrobenzoic acid, pubchem4947, acmc-1cegm | |
C1=C(C(=CC(=C1F)F)[N+](=O)[O-])C(=O)O | |
4219630 | |
99% |
Safety and Handling
- 4,5-Difluoro-2-nitrobenzoic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.