missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-4229181
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
53413-67-5
C9H10BrNO4
276.09
UEKFEYNZISYRRH-UHFFFAOYSA-N
1-bromomethyl-4,5-dimethoxy-2-nitrobenzene, 4,5-dimethoxy-2-nitrobenzyl bromide, 1-bromomethyl-2-nitro-4,5-dimethoxybenzene, bmndmb, benzene, 1-bromomethyl-4,5-dimethoxy-2-nitro, 6-nitroveratryl bromide, acmc-1ao6p, 4,5-dimenoxy-2-nitrobenzyl bromide, 2-nitro-4,5-dimethoxybenzyl bromide, 1-bromomethyl-4,5-dimethoxynitrobenzene
3016812
1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
COC1=C(C=C(C(=C1)CBr)[N+](=O)[O-])OC
This item is not returnable. View return policy
53413-67-5
C9H10BrNO4
276.09
UEKFEYNZISYRRH-UHFFFAOYSA-N
1-bromomethyl-4,5-dimethoxy-2-nitrobenzene, 4,5-dimethoxy-2-nitrobenzyl bromide, 1-bromomethyl-2-nitro-4,5-dimethoxybenzene, bmndmb, benzene, 1-bromomethyl-4,5-dimethoxy-2-nitro, 6-nitroveratryl bromide, acmc-1ao6p, 4,5-dimenoxy-2-nitrobenzyl bromide, 2-nitro-4,5-dimethoxybenzyl bromide, 1-bromomethyl-4,5-dimethoxynitrobenzene
3016812
1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
COC1=C(C=C(C(=C1)CBr)[N+](=O)[O-])OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 130°C to 132°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 276.09
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4, 5-Dimethoxy-2-nitrobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4,5-Dimethoxy-2-nitrobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'