missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine, 95%, Thermo Scientific™

Catalog Number p-7053607
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
150728-13-5
C15H10Cl2N4O2
349.171
MFCD03839838
IZGOBGVYADHVKH-UHFFFAOYSA-N
10545706
4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
COC1=CC=CC=C1OC2=C(N=C(N=C2Cl)C3=NC=CC=N3)Cl
This item is not returnable. View return policy
150728-13-5
C15H10Cl2N4O2
349.171
MFCD03839838
IZGOBGVYADHVKH-UHFFFAOYSA-N
10545706
4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
COC1=CC=CC=C1OC2=C(N=C(N=C2Cl)C3=NC=CC=N3)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 4,6-dichloro-5-2-methoxyphenoxy-2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy-2,2-bipyrimidine, 2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxy-phenoxy-2,2' bipyrimidinyl, 2,2'-bipyrimidine,4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxyphenoxy-2-pyrimidin-2-yl pyrimidine, 4,6-dichloro-5-2-methoxyphenoxy 2,2'-bipyrimidine, 4,6-dichloro-5-o-methoxyphenoxy-2-2-pyrimidinyl pyrimidine, pubchem17752
Formula Weight 349.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'