missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(6-Hydrazino-4-pyrimidinyl)morpholine, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 195.22 |
Percent Purity | 95% |
Chemical Name or Material | 4-(6-Hydrazino-4-pyrimidinyl)morpholine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'