missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3,5-dichloropyridine, 97%, Acros Organics™

Brand:  Acros Organics 367220050

Code : NEW

Additional Details : CAS Number : 22889-78-7 Weight : 0.01000kg


Quantity 5g

 View more versions of this product

Product Code. 15265988

  • 18.70 € / 5g

In stock, ready to ship: 5

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

22889-78-7
163.01
4-amino-3,5-dichloropyridine, 3,5-dichloro-4-aminopyridine, 3,5-dichloro-4-pyridinamine, 4-pyridinamine, 3,5-dichloro, 3,5-dichloro-4-amino pyridine, 3,5-dichloro-4-pyridine amine, 3,5-dichloro-pyridin-4-ylamine, 3,5-dichloro-4-pyridylamine, 3,5-dichloropyridin-4-ylamine, 4-amino-3,5-dichloro pyridine
3,5-dichloropyridin-4-amine
C5H4Cl2N2
ISIQAMHROGZHOV-UHFFFAOYSA-N
89888
C1=C(C(=C(C=N1)Cl)N)Cl

Specifications

4-Amino-3,5-dichloropyridine
1% max.
5g
96% min. (GC)
4-amino-3,5-dichloropyridine, 3,5-dichloro-4-aminopyridine, 3,5-dichloro-4-pyridinamine, 4-pyridinamine, 3,5-dichloro, 3,5-dichloro-4-amino pyridine, 3,5-dichloro-4-pyridine amine, 3,5-dichloro-pyridin-4-ylamine, 3,5-dichloro-4-pyridylamine, 3,5-dichloropyridin-4-ylamine, 4-amino-3,5-dichloro pyridine
C1=C(C(=C(C=N1)Cl)N)Cl
163.01
163.01
Authentic
158.0°C to 160.0°C
22889-78-7
C5H4Cl2N2
ISIQAMHROGZHOV-UHFFFAOYSA-N
3,5-dichloropyridin-4-amine
89888
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Amino-3,5-dichloropyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents