missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorophenylboronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 444500010

Code : 29

Additional Details : CAS Number : 819058-34-9 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11956041

  • 171.00 € / 1g

Estimated Shipment: 28-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

819058-34-9
237.08
4-amino-3-fluorophenylboronic acid pinacol ester, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C12H17BFNO2
AIXGNRNTXUKZLC-UHFFFAOYSA-N
17906172

Specifications

4-Amino-3-fluorophenylboronic acid pinacol ester
101°C to 102°C
819058-34-9
C12H17BFNO2
AIXGNRNTXUKZLC-UHFFFAOYSA-N
17906172
97%
Glass Bottle
1g
96% min. (GC)
4-amino-3-fluorophenylboronic acid pinacol ester, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
237.08
237.08
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Amino-3-fluorophenylboronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents
Certificates