Learn More
4-Aminobenzoic Acid, 99%, ACROS Organics™
Brand: Acros Organics 146210050
Code : 29
Additional Details : CAS Number : 150-13-0 Weight : 5.00000kg
Packaging | Plastic drum |
---|---|
Quantity | 5kg |
Chemical Identifiers
150-13-0 | |
137.138 | |
ALYNCZNDIQEVRV-UHFFFAOYSA-N | |
978 | |
4-aminobenzoic acid |
C7H7NO2 | |
MFCD00007894 | |
p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino | |
CHEBI:30753 | |
C1=CC(=CC=C1C(=O)O)N |
Specifications
4-Aminobenzoic acid | |
Plastic drum | |
186.0°C to 189.0°C | |
150-13-0 | |
100.0 | |
C7H7NO2 | |
MFCD00007894 | |
02,24 | |
p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino | |
C1=CC(=CC=C1C(=O)O)N | |
137.138 | |
CHEBI:30753 | |
Crystalline Powder |
Authentic | |
White to Yellow | |
5kg | |
98.5 | |
99% | |
H2NC6H4CO2H | |
14, 418 | |
15, 418 | |
ALYNCZNDIQEVRV-UHFFFAOYSA-N | |
4-aminobenzoic acid | |
978 | |
137.14 | |
99% |