missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminobenzonitrile, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 83.0°C to 87.0°C |
| Color | Beige to Orange-Yellow or White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | H2NC6H4CN |
| Quantity | 10 g |
| Beilstein | 14, 425 |
| Show More |
Safety and Handling
Product Identifier
- 4-Aminobenzonitrile
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction