missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyl-2-chloropyridine, 95%, Thermo Scientific Chemicals

Catalog Number 11952031
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
144900-57-2
C6H7ClN2
142.59
GGHCWJWUOSNCSK-UHFFFAOYSA-N
2-chloropyridin-4-yl methanamine, 4-pyridinemethanamine, 2-chloro, 2-chloropyridin-4-yl methylamine, 4-aminomethyl-2-chloropyridine, 2-chloro-4-aminomethylpyridine, 2-chloro-4-pyridyl methylamine, 1-2-chloro-4-pyridinyl methanamine, 2-chloro-4-pyridinemethanamine, 1-2-chloropyridin-4-yl methanamine, c-2-chloro-pyridin-4-yl-methylamine
2794167
(2-chloropyridin-4-yl)methanamine
C1=CN=C(C=C1CN)Cl
This item is not returnable. View return policy
144900-57-2
C6H7ClN2
142.59
GGHCWJWUOSNCSK-UHFFFAOYSA-N
2-chloropyridin-4-yl methanamine, 4-pyridinemethanamine, 2-chloro, 2-chloropyridin-4-yl methylamine, 4-aminomethyl-2-chloropyridine, 2-chloro-4-aminomethylpyridine, 2-chloro-4-pyridyl methylamine, 1-2-chloro-4-pyridinyl methanamine, 2-chloro-4-pyridinemethanamine, 1-2-chloropyridin-4-yl methanamine, c-2-chloro-pyridin-4-yl-methylamine
2794167
(2-chloropyridin-4-yl)methanamine
C1=CN=C(C=C1CN)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 142.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Aminomethyl-2-chloropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Aminomethyl-2-chloropyridine, 95%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'