missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetic acid, 98%, ACROS Organics™

Brand:  Acros Organics 104280050

Code : 29

Additional Details : CAS Number : 1197-55-3 Weight : 0.05500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10018661

  • 18.28 € / 5g

Estimated Shipment
01-02-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1197-55-3
151.16
CSEWAUGPAQPMDC-UHFFFAOYSA-N
14533
C1=CC(=CC=C1CC(=O)O)N
C8H9NO2
MFCD00007916
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
2-(4-aminophenyl)acetic acid

Specifications

4-Aminophenylacetic acid
97.5
97.5% min. (HPLC)
H2NC6H4CH2CO2H
MFCD00007916
15, 459
Glass bottle
CSEWAUGPAQPMDC-UHFFFAOYSA-N
2-(4-aminophenyl)acetic acid
14533
Powder
1% max. (K.F.)
196.0°C to 201.0°C
1197-55-3
100.0
C8H9NO2
Authentic
14, 456
4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
Solubility in water: insoluble. Other solubilities: soluble in alkalies, soluble in alcohol
C1=CC(=CC=C1CC(=O)O)N
151.16
151.16
98%
Yellow to Beige
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Aminophenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents