missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-2-(chloromethyl)morpholine, 97%, Thermo Scientific™

Catalog Number 10427262
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
40987-25-5
C12H16ClNO
225.72
MFCD02681886
GVWRZZNYCOTWNN-UHFFFAOYNA-N
4-benzyl-2-chloromethyl morpholine, 4-benzyl-2-chloromethyl-morpholine, 2-chloromethyl-4-benzylmorpholine, morpholine, 2-chloromethyl-4-phenylmethyl, pubchem12932, 3-chloromethyl-4-phenylmethyl morpholine, 2-chloromethyl-4-benzyl morpholine, 2-chloromethyl-4-benzyl-morpholine
2801562
4-benzyl-2-(chloromethyl)morpholine
ClCC1CN(CC2=CC=CC=C2)CCO1
This item is not returnable. View return policy
40987-25-5
C12H16ClNO
225.72
MFCD02681886
GVWRZZNYCOTWNN-UHFFFAOYNA-N
4-benzyl-2-chloromethyl morpholine, 4-benzyl-2-chloromethyl-morpholine, 2-chloromethyl-4-benzylmorpholine, morpholine, 2-chloromethyl-4-phenylmethyl, pubchem12932, 3-chloromethyl-4-phenylmethyl morpholine, 2-chloromethyl-4-benzyl morpholine, 2-chloromethyl-4-benzyl-morpholine
2801562
4-benzyl-2-(chloromethyl)morpholine
ClCC1CN(CC2=CC=CC=C2)CCO1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 225.72
Percent Purity 97%
Chemical Name or Material 4-Benzyl-2-(chloromethyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'