missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylamino-6-chloropyrimidine, 96%, Thermo Scientific™

Product Code. 15406577
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 15406577

Brand: Thermo Scientific Alfa Aesar H33723.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 61667-16-1
Melting Point 116°C to 117°C
Molecular Formula C11H10ClN3
MDL Number MFCD00679879
Quantity 500 mg
Synonym 4-pyrimidinamine, 6-chloro-n-phenylmethyl, n-benzyl-6-chloro-4-pyrimidinamine, benzyl-6-chloro-pyrimidin-4-yl-amine, 4-benzylamino-6-chloropyrimidine, n-benzyl-6-chloropyrimidine-4-amine, 4-pyrimidinamine,6-chloro-n-phenylmethyl
InChI Key XARFPUIPIOPYGA-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CNC2=CC(=NC=N2)Cl
IUPAC Name N-benzyl-6-chloropyrimidin-4-amine
Molecular Weight (g/mol) 219.672
PubChem CID 3771857
Formula Weight 219.67
Percent Purity 96%
Chemical Name or Material 4-Benzylamino-6-chloropyrimidine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.