missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-65536
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4397-53-9
C14H12O2
212.25
MFCD00003387,MFCD01632880
ZVTWZSXLLMNMQC-UHFFFAOYSA-N
4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin
78109
4-phenylmethoxybenzaldehyde
O=CC1=CC=C(OCC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
4397-53-9
C14H12O2
212.25
MFCD00003387,MFCD01632880
ZVTWZSXLLMNMQC-UHFFFAOYSA-N
4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin
78109
4-phenylmethoxybenzaldehyde
O=CC1=CC=C(OCC2=CC=CC=C2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 74°C
Color Cream to Yellow
Boiling Point 197°C to 199°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CHO
Quantity 25 g
Beilstein 08, 73
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and chloroform
Formula Weight 212.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzaldehyde, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'