missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylphenylacetonitrile, 97%, Thermo Scientific™

Product Code. 11374256
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11374256

Brand: Thermo Scientific Alfa Aesar L19332.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101096-72-4
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00060310
InChI Key RNZDBGKQDKFAPR-UHFFFAOYSA-N
Synonym 4-benzylphenylacetonitrile, 2-4-benzylphenyl acetonitrile, acmc-20amog, 4-benzylphenyl acetonitrile, 4-benzyl-l-cyanomethylbenzene, 2-4-benzylphenyl ethanenitrile, benzeneacetonitrile,4-phenylmethyl
PubChem CID 597531
IUPAC Name 2-(4-benzylphenyl)acetonitrile
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)CC#N

Specifications

Melting Point 34°C to 37°C
Boiling Point 145°C to 150°C (0.05 mmHg)
Quantity 250 mg
UN Number UN3439
Beilstein 3258509
Formula Weight 207.28
Percent Purity 97%
Chemical Name or Material 4-Benzylphenylacetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.